missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, MedChemExpress

Product Code. 16459416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459416 Supplier Medchem Express Supplier No. HYY0543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Methylfurfural is a naturally occurring substance, found in cigarette smoke condensate, licorice essential oil, stored dehydrated orange powder, baked potato flour, volatile compounds of roast beef, aroma concentrate of sponge cake. bread and in coffee, tea and cocoa. A flavoring agent.

5-Methylfurfural is a naturally occurring substance, found in cigarette smoke condensate, licorice essential oil, stored dehydrated orange powder, baked potato flour, volatile compounds of roast beef, aroma concentrate of sponge cake. bread and in coffee, tea and cocoa. A flavoring agent.

Specifications

Chemical Name or Material 5-Methylfurfural
CAS 620-02-0
Color Light Yellow to Yellow
Purity Grade Notes Research
Molecular Formula C6H6O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (908.18 mM; Need ultrasonic)
SMILES O=CC1=CC=C(C)O1
Molecular Weight (g/mol) 110.11
Formula Weight 110.11
Percent Purity 97.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.