missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, 98+%

Product Code. 10307770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307770 100 g Glass bottle 100g
10591981 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307770 Supplier Thermo Scientific Acros Supplier No. 159921000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 620-02-0 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Acidity 10mg KOH/g max.
CAS Min % 98.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1000g/mL
Boiling Point 187°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 100 g
Beilstein 17, 289
Specific Gravity 1.1
Solubility Information Solubility in water: 33.3g/L (20 c)
Formula Weight 110.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 5-Methylfurfural, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.