missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5-Methylcytosine Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA130675
This item is not returnable.
View return policy
Description
The use of 7mM NaOH in the IP is crutial. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Methylation of DNA cytosine bases plays important roles in the regulation of gene transcription. It has been estimated that 60 to 90% of the cytosines in CpG dinucleotides are methylated. An increase in methyl-CpG correlates with transcriptional silencing for the whole chromosome especially in developmentally regulated genes. Methylation is believed to lead to transcriptional silencing by multiple mechanisms including alteration of transcription factor binding to promoters and alteration in chromatin structure.
Specifications
| 5-Methylcytosine | |
| Polyclonal | |
| Unconjugated | |
| 5mc; 5-mC; 5-meC; 5-Methyl-cytosine; C5H7NO3 | |
| 5-methyl Cytosine coupled to coupled to keyhole limpet hemocyanin (KLH). | |
| 250 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
| Dot Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3.2 mg/mL | |
| PBS with no preservative | |
| Sheep | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction