missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcytosine, 97%

Product Code. p-7058932
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345827 100 mg 100mg
11355827 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345827 Supplier Thermo Scientific Alfa Aesar Supplier No. 043492.MC
Add to basket
This item is not returnable. View return policy

CAS: 554-01-8 | C5H7N3O | 125.13 g/mol

5-Methylcytosine is used as a marker to protect DNA from being cut by native methylation-sensitive restriction enzymes. It is also employed in the production of 4-amino-5-hydroxy-5-methyl-2-oxo-2,5-dihydro-imidazole-1-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-01-8
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00233537
InChI Key LRSASMSXMSNRBT-UHFFFAOYSA-N
Synonym 5-methylcytosine, cytosine, 5-methyl, 2 1h-pyrimidinone, 4-amino-5-methyl, 5-methylcytosine van, 5 methylcytosine, 5-methyl-cytosine, unii-6r795cqt4h, 4-amino-5-methyl-2-pyrimidinol, 4-amino-5-methylpyrimidin-2 1h-one, cytosine, 5-methyl-van
PubChem CID 65040
ChEBI CHEBI:27551
IUPAC Name 6-amino-5-methyl-1H-pyrimidin-2-one
SMILES CC1=C(N)NC(=O)N=C1

Specifications

Quantity 100 mg
Merck Index 14,6050
Solubility Information Soluble in water.
Formula Weight 125.13
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 5-Methylcytosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.