missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzo[b]thiophene, 97%

Product Code. 11490533
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11490533

Brand: Thermo Scientific Alfa Aesar A12666.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14315-14-1 | C9H8S | 148.22 g/mol

5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14315-14-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00052509
InChI Key DOHZWDWNQFZIKH-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene, benzo b thiophene, 5-methyl, 5-methylbenzothiophene, 5-methyl-benzo b thiophene, 5-methylthianaphthene, 5-methyl-benzothiophene, benzothiophene, 5-mehyl, 5-methyl benzo b thiophene
PubChem CID 84346
IUPAC Name 5-methyl-1-benzothiophene
SMILES CC1=CC=C2SC=CC2=C1

Specifications

Melting Point 32°C to 36°C
Density 1.11
Boiling Point 71°C to 73°C (0.6 mmHg)
Quantity 1 g
Beilstein 109866
Solubility Information Insoluble in water.
Formula Weight 148.23
Percent Purity 97%
Chemical Name or Material 5-Methylbenzo[b]thiophene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.