missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzimidazole, 98%

Product Code. 11448516
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448516 100 g 100g
11428516 5 g 5g
11438516 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448516 Supplier Thermo Scientific Alfa Aesar Supplier No. A15957.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-97-1 | C8H8N2 | 132.166 g/mol

5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

Solubility
Soluble in chloroform (25mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-97-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00010740
InChI Key RWXZXCZBMQPOBF-UHFFFAOYSA-N
Synonym 5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole, 1h-benzimidazole, 5-methyl, 5-methyl-1h-benzimidazole, benzimidazole, 5-methyl, ethyl 4-cyanophenylalaninate, 1h-benzimidazole, 6-methyl, 5-methyl-1h-1,3-benzodiazole, 6-methyl-1h-1,3-benzodiazole, 5mb
PubChem CID 11979
ChEBI CHEBI:40205
IUPAC Name 6-methyl-1H-benzimidazole
SMILES CC1=CC2=C(C=C1)N=CN2

Specifications

Melting Point 112°C to 117°C
Boiling Point 169°C to 172°C (1 mmHg)
Quantity 100 g
Beilstein 2628
Solubility Information Soluble in chloroform (25mg/ml).
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 5-Methylbenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.