missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl Cytosine Hydrochloride, TRC

Product Code. 30471652
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471652 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471652 Supplier Toronto Research Chemicals Supplier No. TRCM2947001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methylcytosine Hydrochloride
CAS 58366-64-6
Molecular Formula C5 H7 N3 O . Cl H
Quantity 1 g
Synonym 2(1H)-Pyrimidinone, 6-amino-5-methyl-, hydrochloride (1:1), 2(1H)-Pyrimidinone, 4-amino-5-methyl-, monohydrochloride (9CI), Cytosine, 5-methyl-, hydrochloride (6CI), 5-Methylcytosine hydrochloride, 6-Amino-5-methyl-2(1H)-pyrimidinone hydrochloride, 4-Amino-2-hydroxy-5-methylpyrimidine hydrochloride, 4-Amino-5-methyl-2(1H)-pyrimidinone hydrochloride, 5-Methyl cytosine hydrochloride
InChI Formula InChI=1S/C5H7N3O.ClH/c1-3-2-7-5(9)8-4(3)6;/h2H,1H3,(H3,6,7,8,9);1H
SMILES Cl.CC1=CNC(=O)N=C1N
IUPAC Name 4-amino-5-methyl-1H-pyrimidin-2-one;hydrochloride
Molecular Weight (g/mol) 161.59
Formula Weight 161.0356
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.