missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl Chrysene-d3 (1mg/mL In Dichloromethane), TRC
Specifications
Specifications
| Solvent or Matrix | Dichloromethane |
| Molecular Formula | C19H11D3 |
| Quantity | 1 mL |
| Synonym | 5-Methylchrysene-d3, NSC 407620-d3 |
| InChI Formula | InChI=1S/C19H14/c1-13-12-15-7-3-4-8-16(15)18-11-10-14-6-2-5-9-17(14)19(13)18/h2-12H,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])C1=CC2=CC=CC=C2C3=C1C(C=CC=C4)=C4C=C3 |
| Molecular Weight (g/mol) | 245.33 |
| Concentration or Composition (by Analyte or Components) | 5-Methyl Chrysene-d3 1 mg/mL |
| Chemical Name or Material | 5-Methyl Chrysene-d3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?