missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-5-phenylhydantoin, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-5-phenyl-2,4-imidazolidinedione |
| CAS | 6843-49-8 |
| Molecular Formula | C10 H10 N2 O2 |
| Quantity | 25 mg |
| Synonym | Hydantoin, 5-methyl-5-phenyl- (6CI,7CI,8CI), 5-Methyl-5-phenyl-2,4-imidazolidinedione, (±)-5-Methyl-5-phenylhydantoin, 5-Methyl-5-phenylhydantoin, 5-Methyl-5-phenylimidazolidine-2,4-dione, NSC 14839 |
| InChI Formula | InChI=1S/C10H10N2O2/c1-10(7-5-3-2-4-6-7)8(13)11-9(14)12-10/h2-6H,1H3,(H2,11,12,13,14) |
| SMILES | CC1(NC(=O)NC1=O)c2ccccc2 |
| IUPAC Name | 5-methyl-5-phenylimidazolidine-2,4-dione |
| Molecular Weight (g/mol) | 190.2 |
| Formula Weight | 190.0742 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction