missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-4-phenyl-2-(2-pyridyl)thiazole, 97%, Thermo Scientific™

Catalog Number 15468227
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1217863-27-8
C15H12N2S
252.335
MFCD16251563
ZTYJKDZBAFCCRT-UHFFFAOYSA-N
5-methyl-4-phenyl-2-2-pyridyl thiazole, 2-5-methyl-4-phenyl-1,3-thiazol-2-yl pyridine, 5-methyl-4-phenyl-2-pyridin-2-yl thiazole
73012868
5-methyl-4-phenyl-2-pyridin-2-yl-1,3-thiazole
CC1=C(N=C(S1)C2=CC=CC=N2)C3=CC=CC=C3
This item is not returnable. View return policy
1217863-27-8
C15H12N2S
252.335
MFCD16251563
ZTYJKDZBAFCCRT-UHFFFAOYSA-N
5-methyl-4-phenyl-2-2-pyridyl thiazole, 2-5-methyl-4-phenyl-1,3-thiazol-2-yl pyridine, 5-methyl-4-phenyl-2-pyridin-2-yl thiazole
73012868
5-methyl-4-phenyl-2-pyridin-2-yl-1,3-thiazole
CC1=C(N=C(S1)C2=CC=CC=N2)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 84°C to 88°C
Quantity 1 g
Formula Weight 252.33
Percent Purity 97%
Chemical Name or Material 5-Methyl-4-phenyl-2-(2-pyridyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'