missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-4-isoxazolecarbonyl chloride, 97%, Thermo Scientific™

Product Code. 10371573
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 67305-24-2
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.542
InChI Key ZKAQPVQEYCFRTK-UHFFFAOYSA-N
Synonym 5-methyl-4-isoxazolecarbonyl chloride, 5-methylisoxazole-4-carbonyl chloride, 5-methyl-4-isoxazolecarbonylchloride, 4-isoxazolecarbonyl chloride, 5-methyl, 4-isoxazolecarbonylchloride, 5-methyl, 5-methyl-isoxazole-4-carbonyl chloride, acmc-209nyg, methyl-4-isoxazolecarbonyl chloride, 4-isoxazolecarbonylchloride,5-methyl
PubChem CID 2759917
IUPAC Name 5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C=NO1)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 145.54
Percent Purity 97%
Chemical Name or Material 5-Methyl-4-isoxazolecarbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'