missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-trifluoromethyl-1H-pyrazole, 98%, Thermo Scientific™

Product Code. 15411459
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411459 1 g 1g
15431459 250 mg 250mg
15421459 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411459 Supplier Thermo Scientific Alfa Aesar Supplier No. H64308.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10010-93-2
Molecular Formula C5H5F3N2
Molecular Weight (g/mol) 150.104
MDL Number MFCD00156058
InChI Key DLCHCAYDSKIFIN-UHFFFAOYSA-N
Synonym 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
PubChem CID 139077
IUPAC Name 5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=CC(=NN1)C(F)(F)F

Specifications

Melting Point 88°C to 90°C
Color White
Quantity 1 g
Beilstein 743044
Formula Weight 150.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Methyl-3-trifluoromethyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.