missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carboxylic Acid, Mikromol™

Product Code. 30049463
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049463 100 mg 100mg
30052735 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049463 Supplier Mikromol Supplier No. MM0686.10
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methyl-3-phenylisoxazole-4-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carboxylic acid
CAS 1136-45-4
Analyte or Component Names 5-Methyl-3-phenylisoxazole-4-carboxylic acid
Shipping Condition Room Temperature
Molecular Formula C11 H9 N O3
Certifications/Compliance ISO 17025
Synonym 5-Methyl-3-phenylisoxazole-4-carboxylic acid, Oxacillin Related Compound C, Oxacillin Sodium Monohydrate Impurity C, Oxacillin Impurity C
InChI Formula InChI=1S/C11H9NO3/c1-7-9(11(13)14)10(12-15-7)8-5-3-2-4-6-8/h2-6H,1H3,(H,13,14)
SMILES Cc1onc(c2ccccc2)c1C(=O)O
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol) 203.19
Quantity 100 mg
Formula Weight 203.0582 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.