missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-4-isoxazolecarboxylic Acid, TRC

Product Code. 30481925
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481925 Supplier Toronto Research Chemicals Supplier No. TRCM2277351G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1136-45-4
Molecular Formula C11 H9 N O3
Quantity 1 g
Synonym 5-Methyl-3-phenylisoxazole-4-carboxylic acid
InChI Formula InChI=1S/C11H9NO3/c1-7-9(11(13)14)10(12-15-7)8-5-3-2-4-6-8/h2-6H,1H3,(H,13,14)
SMILES Cc1onc(c2ccccc2)c1C(=O)O
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol) 203.19
Formula Weight 203.0582
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.