Learn More
Thermo Scientific Maybridge 5-Methyl-3-isoxazolecarbaldehyde, 5g, 97%
Brand: Thermo Scientific Maybridge CC10404DE
5-Methyl-3-isoxazolecarbaldehyde, 5g, 97%
Code : 31
Additional Details : CAS Number : 62254-74-4 Weight : 0.00500kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III