missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15458065
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458065 250 mg 250mg
15445077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458065 Supplier Thermo Scientific Alfa Aesar Supplier No. H32357.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1017778-27-6
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD09832298
InChI Key UMEQVNCETMDPES-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl phenylacetic acid, 2-5-methyl-2-trifluoromethyl phenyl acetic acid, 5-methyl-2-trifluoromethyl phenyl acetic acid
PubChem CID 46737529
IUPAC Name 2-[5-methyl-2-(trifluoromethyl)phenyl]acetic acid
SMILES CC1=CC(=C(C=C1)C(F)(F)F)CC(=O)O

Spécification

Melting Point 117°C to 120°C
Quantity 250 mg
Formula Weight 218.18
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)phenylacetic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.