missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11411310
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411310 5 g 5g
11401310 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411310 Supplier Thermo Scientific Alfa Aesar Supplier No. H26377.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120985-68-4
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD06660291
InChI Key YYEMDMKMHBXYPV-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl benzoic acid, benzoic acid, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl benzoicacid, 5-methyl-2-trifluoromethyl-benzoic acid
PubChem CID 14203325
IUPAC Name 5-methyl-2-(trifluoromethyl)benzoic acid
SMILES CC1=CC(=C(C=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 90°C to 94°C
Quantity 5 g
Solubility Information Soluble in alcohol.
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.