missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-thiophenecarboxaldehyde, 98%

Product Code. 10390894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390894 50 mL Glass bottle 50mL
10467900 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10390894 Supplier Thermo Scientific Acros Supplier No. 127870500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13679-70-4 | C6H6OS | 126.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-70-4
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.18
MDL Number MFCD00005434
InChI Key VAUMDUIUEPIGHM-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde
PubChem CID 61663
IUPAC Name 5-methylthiophene-2-carbaldehyde
SMILES CC1=CC=C(S1)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1800g/mL
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5800 to 1.584
Quantity 50 mL
Beilstein 17, I, 151
Specific Gravity 1.18
Formula Weight 126.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-thiophenecarboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.