missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-pyrazinecarboxylic acid, 99%, Thermo Scientific™

Product Code. 10636481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636481 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10636481 Supplier Thermo Scientific Acros Supplier No. 297010050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5521-55-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 158.0°C to 160.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H6N2O2
MDL Number MFCD00068241
Quantity 5 g
Synonym 5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
InChI Key RBYJWCRKFLGNDB-UHFFFAOYSA-N
SMILES CC1=NC=C(N=C1)C(=O)O
IUPAC Name 5-methylpyrazine-2-carboxylic acid
Molecular Weight (g/mol) 138.13
PubChem CID 122831
Formula Weight 138.13
Percent Purity 99%
Physical Form Amorphous Powder
Chemical Name or Material 5-Methyl-2-pyrazinecarboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.