missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-3-furyl)methanol, 95%, Thermo Scientific™

Product Code. 10397344
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397344 1 g Amber Glass Bottle 1g
10708075 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10397344 Supplier Thermo Scientific Maybridge Supplier No. CC49709DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 183210-33-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD08435905
InChI Key VZFFMWAZPODJRX-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furyl methanol, 5-methyl-2-phenylfuran-3-yl methanol, 3-furanmethanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methan-1-ol
PubChem CID 15269738
IUPAC Name (5-methyl-2-phenylfuran-3-yl)methanol
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material (5-Methyl-2-phenyl-3-furyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.