missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furanmethanamine, 98%

Product Code. 10093490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10093490 25 g Glass bottle 25g
10159322 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10093490 Supplier Thermo Scientific Acros Supplier No. 268940250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14003-16-8 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14003-16-8
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00143471
InChI Key YSEAGSCGERFGBL-UHFFFAOYSA-N
Synonym 5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz
PubChem CID 2724683
IUPAC Name (5-methylfuran-2-yl)methanamine
SMILES CC1=CC=C(CN)O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -22°C
Color Yellow
Density 1.0100g/mL
Boiling Point 57°C (13.0 Mbar)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4860 to 1.488
Quantity 25 g
Specific Gravity 1.01
Solubility Information Solubility in water: >1000g/L (20°C)
Formula Weight 111.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furanmethanamine, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5-Methyl-2-furanmethanamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.