missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2'-deoxycytidine, MP Biomedicals™

Product Code. p-4405881 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg

Product Code. 11486861

Brand: MP Biomedicals 0219888310

Please to purchase this item. Need a web account? Register with us today!

5-Methyl-2'-deoxycytidine, White Powder

  • 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
  • It has been used in epigenetics research to investigate the dynamics of DNA methylation pattern in the control of gene expression.
  • A cytidine analog and an isostere of thymidine.
  • 5-Methyl-2'-deoxycytidine is a pyrimidine nucleoside that when incorporated into single-stranded DNA can act in cis to signal de novo DNA methylation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-07-3
MDL Number MFCD00006549
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N
PubChem CID 440055
ChEBI CHEBI:47876
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
SMILES CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O

Specifications

Melting Point 217°C to 219°C (Literature)
Color White
Assay Percent Range >99.5%
Molecular Formula C10H15N5O3
Refractive Index 1.694 (lit.)
Quantity 10 mg
Synonym 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
Solubility Information Soluble in 1 N NaOH.
Optical Rotation [α]25/D +63°, c = 1 in 1 M NaOH (lit.)
Molecular Weight (g/mol) 241.2
Formula Weight 241.2
Vapor Pressure 8.85E-12mmHg at 25°C (lit.)
Physical Form Powder
Chemical Name or Material 5-Methyl-2'-deoxycytidine
Grade 99.6 area% (HPLC)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.