missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2'-deoxycytidine, MP Biomedicals™

Product Code. 11475266 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
This item is not returnable. View return policy

Product Code. 11475266

Brand: MP Biomedicals 0219888380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Methyl-2'-deoxycytidine, White Powder

  • 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
  • It has been used in epigenetics research to investigate the dynamics of DNA methylation pattern in the control of gene expression.
  • A cytidine analog and an isostere of thymidine.
  • 5-Methyl-2'-deoxycytidine is a pyrimidine nucleoside that when incorporated into single-stranded DNA can act in cis to signal de novo DNA methylation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 838-07-3
MDL Number MFCD00006549
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N

Specifications

Melting Point 217°C to 219°C (literature)
Color White
Molecular Formula C10H15N3O4
Refractive Index 1.694 (lit.)
Quantity 100 mg
Synonym 2'-deoxy-5-methyl-cytidine
Solubility Information Soluble in 1 N NaOH.
Optical Rotation [α]25/D +63°, c = 1 in 1 M NaOH (lit.)
Molecular Weight (g/mol) 241.25
Vapor Pressure 8.85E-12mmHg at 25°C (lit.)
Physical Form Powder
Grade 99.6 area% (HPLC)
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 5-Methyl-2'-deoxycytidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.