Learn More
Alfa Aesar™ 5-Methyl-2-(3-nitrophenyl)-3-pyrazolinone, 98%
Brand: Alfa Aesar™ H63300.03
Code : NEW
Additional Details : CAS Number : 119-16-4 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
5-Methyl-2-(3-nitrophenyl)-3-pyrazolinone | |
121.21 | |
3-Methyl-1-(3-nitrophenyl)-5-pyrazolone; 5-Methyl-2-(3-nitrophenyl)-1H-pyrazol-3(2H)-one |
119-16-4 | |
MFCD00044057 |
Specifications
5-Methyl-2-(3-nitrophenyl)-3-pyrazolinone | |
119-16-4 | |
98% | |
MFCD00044057 |
1g | |
Solid | |
121.21 | |
3-Methyl-1-(3-nitrophenyl)-5-pyrazolone; 5-Methyl-2-(3-nitrophenyl)-1H-pyrazol-3(2H)-one |