missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-tetrazole, 97%

Product Code. 15406728
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15406728

Brand: Thermo Scientific Alfa Aesar H60248.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4076-36-2 | C2H4N4 | 84.082 g/mol

5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4076-36-2
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00129971
InChI Key XZGLNCKSNVGDNX-UHFFFAOYSA-N
Synonym 5-methyltetrazole, 5-methyl-1h-tetrazole, 5-methyl tetrazole, 1h-tetrazole, 5-methyl, 5-methyl-1h-tertazole, tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetrazole, 5-methyl-2h-1,2,3,4-tetrazole, 2h-tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetraazole
PubChem CID 138107
IUPAC Name 5-methyl-2H-tetrazole
SMILES CC1=NNN=N1

Specifications

Melting Point 142°C to 146°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 84.08
Percent Purity 97%
Chemical Name or Material 5-Methyl-1H-tetrazole
compliance-icons
Product Identifier
  • 5-Methyl-1H-tetrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 5-Methyl-1H-tetrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.