missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole, 98%, Thermo Scientific™

Product Code. 10276190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10276190 50 g Glass bottle 50g
10367460 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10276190 Supplier Thermo Scientific Acros Supplier No. 188000500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-85-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005702
InChI Key LRUDIIUSNGCQKF-UHFFFAOYSA-N
Synonym 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
PubChem CID 8705
ChEBI CHEBI:83455
IUPAC Name 5-methyl-2H-benzotriazole
SMILES CC1=CC2=NNN=C2C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80.0°C to 84.0°C
Color Cream to Beige
Boiling Point 210.0°C to 212.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 26, 58
Solubility Information Solubility in water: 6.0g/L (25 c). Other solubilities: soluble in acids and alkalies,soluble in methanol and ethanol
Formula Weight 133.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1H-benzotriazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methyl-1H-benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.