missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2703022
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
91138-00-0
C11H10N2O2
202.213
USSMIQWDLWJQDQ-UHFFFAOYSA-N
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
145221
5-methyl-1-phenylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
This item is not returnable. View return policy
91138-00-0
C11H10N2O2
202.213
USSMIQWDLWJQDQ-UHFFFAOYSA-N
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
145221
5-methyl-1-phenylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'