missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 5-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-phenyl-1h-pyrazole-4-carboxaldehyde, acmc-20am2d, 1h-pyrazole-4-carboxaldehyde, 5-methyl-1-phenyl, 1h-pyrazole-4-carboxaldehyde,5-methyl-1-phenyl |
Formula Weight | 186.21 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'