missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 95%, Thermo Scientific™

Product Code. 11936221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936221 5 g 5g
11926221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936221 Supplier Thermo Scientific Acros Supplier No. 439760050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 894779-67-0
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.24
InChI Key GRLXUIRJMGNTQO-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine, 1h-pyrazole-4-methanamine, 5-methyl-1-phenyl, 5-methyl-1-phenylpyrazol-4-yl methanamine, 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine
PubChem CID 28472963
IUPAC Name (5-methyl-1-phenylpyrazol-4-yl)methanamine
SMILES CC1=C(C=NN1C2=CC=CC=C2)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 187.24
Percent Purity 95%
Physical Form Liquid or Powder
Chemical Name or Material 5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
compliance-icons compliance-icons
Product Identifier
  • 5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.