missing translation for 'onlineSavingsMsg'
Learn More

5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10193084
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193084 5 g 5g
10262592 250 mg 250mg
10049281 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193084 Supplier Thermo Scientific Maybridge Supplier No. CD04550DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499771-21-0
Molecular Formula C11H8F3N3O2
Molecular Weight (g/mol) 271.199
InChI Key IRJLZHQBWAUKGA-UHFFFAOYSA-N
Synonym 5-methyl-1-3-trifluoromethyl phenyl-1h-1,2,3-triazole-4-carboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl-1,2,3-triazole-4-carboxylic acid, maybridge1_002663, 5-methyl-1-3-trifluoromethyl phenyl-4-triazolecarboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl triazole-4-carboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl-1h-1,2,3-triazole-4carboxylic acid
PubChem CID 2804089
IUPAC Name 5-methyl-1-[3-(trifluoromethyl)phenyl]triazole-4-carboxylic acid
SMILES CC1=C(N=NN1C2=CC=CC(=C2)C(F)(F)F)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 271.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-[3-(trifluoromethyl)phenyl]-1H-1, 2, 3-triazole-4-carboxylic acid
compliance-icons
Product Identifier
  • 5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.