missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate, 99%, Thermo Scientific™

Product Code. 11499280
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499280 250 mg 250mg
11409290 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499280 Supplier Thermo Scientific Alfa Aesar Supplier No. H50162.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187931-57-2
Molecular Formula C6H9N3O5
Molecular Weight (g/mol) 203.15
MDL Number MFCD07781053
InChI Key BQNHEXDOLOGXGU-UHFFFAOYSA-N
Synonym 5-methyl-1,3,4-oxadiazol-2-yl methanamine oxalate, c-5-methyl-1,3,4 oxadiazol-2-yl-methylamine oxalate, 5-methyl-1,3,4-oxadiazole-2-methylamine oxalate, c4h7n3o.c2h2o4, c-5-methyl-1,3,4 oxadiazol-2yl-methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methanamine; oxalic acid
PubChem CID 53445292
IUPAC Name (5-methyl-1,3,4-oxadiazol-2-yl)methanamine;oxalic acid
SMILES OC(=O)C(O)=O.CC1=NN=C(CN)O1

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 203.15
Percent Purity 99%
Chemical Name or Material 5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.