missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. p-7047384
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11319625

Brand: Thermo Scientific Alfa Aesar L19070.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-83-1
Molecular Formula C11H15ClN2O
Molecular Weight (g/mol) 226.704
MDL Number MFCD00012684
InChI Key TXVAYRSEKRMEIF-UHFFFAOYSA-N
Synonym 5-methoxytryptamine hydrochloride, mexamine hydrochloride, o-methylserotonin hydrochloride, 2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methoxyindole hydrochloride, unii-4265ib6qek, 5-methoxy-1h-indole-3-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride, 5-methoxy tryptamine hcl, 5 methoxytryptamine hydrochloride
PubChem CID 6198
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES COC1=CC2=C(C=C1)NC=C2CCN.Cl

Specifications

Melting Point 245°C to 250°C (decomposition)
Odor Odorless
Quantity 250 mg
Beilstein 3717598
Sensitivity Hygroscopic
Merck Index 14,6002
Formula Weight 226.71
Percent Purity ≥98%
Chemical Name or Material 5-Methoxytryptamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.