missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10031440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031440 1 g Glass bottle 1g
10386670 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031440 Supplier Thermo Scientific Acros Supplier No. 126130010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 66-83-1
Molecular Formula C11H14N2O·HCl
Molecular Weight (g/mol) 226.71
MDL Number MFCD00012684
InChI Key TXVAYRSEKRMEIF-UHFFFAOYSA-N
Synonym 5-methoxytryptamine hydrochloride, mexamine hydrochloride, o-methylserotonin hydrochloride, 2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methoxyindole hydrochloride, unii-4265ib6qek, 5-methoxy-1h-indole-3-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride, 5-methoxy tryptamine hcl, 5 methoxytryptamine hydrochloride
PubChem CID 6198
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES COC1=CC2=C(C=C1)NC=C2CCN.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246.0°C to 248.0°C
Color Beige to White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Merck Index 14, 6002
Formula Weight 226.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxytryptamine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxytryptamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.