missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxyresorcinol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 5-Methoxyresorcinol |
| CAS | 2174-64-3 |
| Molecular Formula | C7 H8 O3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C7H8O3/c1-10-7-3-5(8)2-6(9)4-7/h2-4,8-9H,1H3 |
| SMILES | COc1cc(O)cc(O)c1 |
| IUPAC Name | 5-methoxybenzene-1,3-diol |
| Molecular Weight (g/mol) | 140.137 |
| Formula Weight | 140.047 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction