missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyresorcinol, 95%, Thermo Scientific Chemicals

Product Code. 15488588
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2174-64-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00002285
InChI Key HDVRLUFGYQYLFJ-UHFFFAOYSA-N
Synonym 5-methoxyresorcinol, flamenol, 3,5-dihydroxyanisole, phloroglucinol monomethyl ether, 1,3-benzenediol, 5-methoxy, 5-methoxy-benzene-1,3-diol, unii-6201e0jif3, dsstox_cid_26525, dsstox_rid_81690, dsstox_gsid_46525
PubChem CID 71648
IUPAC Name 5-methoxybenzene-1,3-diol
SMILES COC1=CC(=CC(=C1)O)O

Specifications

Melting Point 78°C to 80°C
Color Yellow
Boiling Point 188°C to 189°C (12 mmHg)
Quantity 1 g
Beilstein 1423578
Solubility Information Soluble in water.
Formula Weight 140.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Methoxyresorcinol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'