missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypsoralen, 98%

Product Code. 15559533 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15559533 1 g 1g
15569533 5 g 5g
15579533 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15559533 Supplier Thermo Scientific Acros Supplier No. 460660010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 484-20-8 | C12H8O4 | 216.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-20-8
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
InChI Key BGEBZHIAGXMEMV-UHFFFAOYSA-N
Synonym bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
PubChem CID 2355
ChEBI CHEBI:18293
IUPAC Name 4-methoxyfuro[3,2-g]chromen-7-one
SMILES COC1=C2C=CC(=O)OC2=CC3=C1C=CO3

Specifications

Melting Point 190.0°C to 193.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.19
Percent Purity 98%
Chemical Name or Material 5-Methoxypsoralen
compliance-icons
Product Identifier
  • 5-Methoxypsoralen
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.