missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxylic acid, 97%

Product Code. 15472029
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15472029

Brand: Thermo Scientific Alfa Aesar H64722.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10242-01-0 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10242-01-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD03265451
InChI Key RVVSEZGJCOAUED-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
PubChem CID 259191
IUPAC Name 5-methoxy-1H-indole-3-carboxylic acid
SMILES COC1=CC2=C(C=C1)NC=C2C(=O)O

Specifications

Color Red
Quantity 250 mg
Beilstein 475711
Formula Weight 191.19
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 5-Methoxyindole-3-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.