missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic acid ethyl ester, 98%, Thermo Scientific™

Product Code. 11956691 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11956691

Brand: Thermo Scientific Acros 440170100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4792-58-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 152°C to 155°C
Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H13NO3
Quantity 10 g
Synonym ethyl 5-methoxyindole-2-carboxylate, 5-methoxy-1h-indole-2-carboxylic acid ethyl ester, ethyl5-methoxyindole-2-carboxylate, ethyl 5-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylic acid, 5-methoxy-, ethyl ester, unii-d63u367gnz, 5-methoxyindole-2-carboxylic acid ethyl ester, methoxy-5 indole carboxylate d'ethyle-2, ethyl 5-methoxyl-1h-indole-2-carboxylate, indole-2-carboxylic acid, 5-methoxy-, ethyl ester
InChI Key NPIUAXNFAUGNHP-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OC
IUPAC Name ethyl 5-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol) 219.24
PubChem CID 20926
Formula Weight 219.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid ethyl ester
Show More Show Less
Product Identifier
  • 5-Methoxyindole-2-carboxylic acid ethyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.