missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic acid, 99%, Thermo Scientific™

Product Code. 10336720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336720 1 g Glass bottle 1g
10695552 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336720 Supplier Thermo Scientific Acros Supplier No. 125900010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4382-54-1
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005614
InChI Key YEBJVSLNUMZXRJ-UHFFFAOYSA-N
Synonym 5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
PubChem CID 20401
IUPAC Name 5-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 197°C to 201°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 22, II, 169
Formula Weight 191.19
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.