missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic acid, 99%, Thermo Scientific™

Catalog Number p-157840
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4382-54-1
C10H9NO3
191.19
MFCD00005614
YEBJVSLNUMZXRJ-UHFFFAOYSA-N
5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
20401
5-methoxy-1H-indole-2-carboxylic acid
COC1=CC=C2NC(=CC2=C1)C(O)=O
This item is not returnable. View return policy
4382-54-1
C10H9NO3
191.19
MFCD00005614
YEBJVSLNUMZXRJ-UHFFFAOYSA-N
5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
20401
5-methoxy-1H-indole-2-carboxylic acid
COC1=CC=C2NC(=CC2=C1)C(O)=O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 197°C to 201°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 22, II, 169
Formula Weight 191.19
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'