missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-alpha-methyltryptamine Hydrochloride (5-MeO-AMT HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30458397
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458397 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458397 Supplier LoGiCal Supplier No. LGCAMP1422.0901

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methoxy-alpha-methyltryptamine Hydrochloride
CAS 1016-44-0
Analyte or Component Names 5-Methoxy-alpha-methyltryptamine Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H16 N2 O . Cl H
Concentration 1 mg/ml
Certifications/Compliance ISO 17034
Synonym 5-Methoxy-α-methyl-1H-indole-3-ethanamine Hydrochloride (1:1), 5-Methoxy-α-methyltryptamine Hydrochloride, 3-(2-Aminopropyl)-5-methoxyindole Hydrochloride (6CI,7CI,8CI)
InChI Formula InChI=1S/C12H16N2O.ClH/c1-8(13)5-9-7-14-12-4-3-10(15-2)6-11(9)12;/h3-4,6-8,14H,5,13H2,1-2H3;1H
Solution Type Single Solution
SMILES Cl.COc1ccc2[nH]cc(CC(C)N)c2c1
IUPAC Name 1-(5-methoxy-1H-indol-3-yl)propan-2-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 240.729
Quantity 1 mL
Formula Weight 240.103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.