missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7031493
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
216766-13-1
C7H6F3NO
177.13
MFCD09864699
MRHJZBSCOLVMGA-UHFFFAOYSA-N
5-methoxy-2-trifluoromethyl pyridine, 5-methoxy-2-trifluoromethyl-pyridine, pubchem23560, 3-methoxy-6-trifluoromethylpyridine, 3-methoxy-6-trifluoromethyl pyridine, pyridine,5-methoxy-2-trifluoromethyl
22000196
5-methoxy-2-(trifluoromethyl)pyridine
COC1=CN=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
216766-13-1
C7H6F3NO
177.13
MFCD09864699
MRHJZBSCOLVMGA-UHFFFAOYSA-N
5-methoxy-2-trifluoromethyl pyridine, 5-methoxy-2-trifluoromethyl-pyridine, pubchem23560, 3-methoxy-6-trifluoromethylpyridine, 3-methoxy-6-trifluoromethyl pyridine, pyridine,5-methoxy-2-trifluoromethyl
22000196
5-methoxy-2-(trifluoromethyl)pyridine
COC1=CN=C(C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 177.13
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'