missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitroaniline, 98%

Product Code. 15486947
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486947 1 g 1g
15496947 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486947 Supplier Thermo Scientific Alfa Aesar Supplier No. H34046.03
 Restricted Item
Estimated Shipment: 13-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 16133-49-6 | C7H8N2O3 | 168.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16133-49-6
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00179573
InChI Key QEHVRGACCVLLNN-UHFFFAOYSA-N
Synonym 5-methoxy-2-nitrophenylamine, 3-amino-4-nitroanisole, 5methoxy-2-nitroaniline, 5-methoxy-2-nitrobenzenamine, 3-amino-4-nitro-anisol, 2-nitro-5-methoxyaniline, 5-methoxy-2-nitro-aniline, benzenamine, 5-methoxy-2-nitro, benzenamine,5-methoxy-2-nitro, 5-methoxy-2-nitro-phenylamine
PubChem CID 85300
IUPAC Name 5-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)[N+](=O)[O-])N

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 5-Methoxy-2-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.