missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylindole, 99+%

Product Code. 11459277
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459277 5 g 5g
11449277 1 g 1g
11469277 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459277 Supplier Thermo Scientific Alfa Aesar Supplier No. B21348.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1076-74-0 | C10H11NO | 161.20 g/mol

It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.

Solubility
Soluble in methanol (very faint turbidity.)

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076-74-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005620
InChI Key VSWGLJOQFUMFOQ-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole
PubChem CID 70642
IUPAC Name 5-methoxy-2-methyl-1H-indole
SMILES COC1=CC=C2NC(C)=CC2=C1

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in methanol (very faint turbidity.)
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material 5-Methoxy-2-methylindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.