missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylbenzothiazole 98%, Thermo Scientific™

Product Code. 10510964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10510964 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10510964 Supplier Thermo Scientific Acros Supplier No. 413840010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2941-69-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C9H9NOS
MDL Number MFCD00082664
Quantity 1g
Synonym 5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
InChI Key SAQMNBWVOKYKPZ-UHFFFAOYSA-N
SMILES COC1=CC=C2SC(C)=NC2=C1
IUPAC Name 5-methoxy-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol) 179.24
PubChem CID 76252
Formula Weight 179.24
Percent Purity 98%
Chemical Name or Material 5-Methoxy-2-methylbenzothiazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.