missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylaniline, 97%

Product Code. 15496648
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496648 1 g 1g
15406658 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496648 Supplier Thermo Scientific Alfa Aesar Supplier No. H60177.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50868-72-9 | C8H11NO | 137.18 g/mol

5-Methoxy-2-methylaniline was used in the preparation of N-(5-methoxy-2-methylphenyl)acetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxy-2-methylaniline was used in the preparation of N-(5-methoxy-2-methylphenyl)acetamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50868-72-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075057
InChI Key RPJXLEZOFUNGNZ-UHFFFAOYSA-N
Synonym 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic
PubChem CID 99500
IUPAC Name 5-methoxy-2-methylaniline
SMILES COC1=CC=C(C)C(N)=C1

Specifications

Melting Point 43°C to 46°C
Flash Point 113°C (235°F)
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.