missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-mercaptobenzimidazole, TRC

Product Code. 30010347
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010347 5 g 5g
30003676 10 g 10g
17984695 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010347 Supplier Toronto Research Chemicals Supplier No. TRCM2627605G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37052-78-1
Molecular Formula C8 H8 N2 O S
Molecular Weight (g/mol) 180.23
Synonym 5-Methoxy-1H-benzimidazole-2-thiol, Esomeprazole Magnesium Trihydrate Imp. A (EP), Omeprazole Imp. A (EP), 2H-Benzimidazole-2-thione, 1,3-dihydro-5-methoxy- (9CI, ACI), 1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione (ACI), 1,3-Dihydro-5-methoxybenzimidazole-2-thione, 2-Mercapto-5-methoxy-1H-benzimidazole, 2-Mercapto-5-methoxybenzimidazole, 2-Thio-5-methoxy-1H-benzimidazole, 5-Methoxy-1H-benzo[d]imidazole-2(3H)-thione, 5-Methoxy-1H-benzoimidazole-2-thiol, 5-Methoxy-2-benzimidazolethiol, 5-Methoxy-2-mercapto-1H-benzimidazole, 5-Methoxy-2-mercaptobenzimidazole
IUPAC Name 5-methoxy-1H-benzimidazole-2-thiol
SMILES COc1ccc2[nH]c(S)nc2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4H,1H3,(H2,9,10,12)
Formula Weight 180.0357
Percent Purity >95%
Chemical Name or Material 5-Methoxy-1H-benzimidazole-2-thiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.