missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-indanone, 98%

Product Code. 10042390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10042390 10 g 10g
10501441 5 g 5g
10678894 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10042390 Supplier Thermo Scientific Acros Supplier No. 179670100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5111-70-6 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5111-70-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00003789
InChI Key QOPRWBRNMPANKN-UHFFFAOYSA-N
Synonym 5-methoxy-1-indanone, 5-methoxyindan-1-one, 5-methoxy-2,3-dihydro-1h-inden-1-one, 5-methoxyindanone, 2,3-dihydro-5-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-5-methoxy, 5-methoxy indanone, 5-methoxyhydrindone, pubchem2031, 1-oxo-5-methoxyindane
PubChem CID 78787
IUPAC Name 5-methoxy-2,3-dihydroinden-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 107.0°C to 111.0°C
Color Brown-Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 08, II, 152
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-1-indanone
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxy-1-indanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.