missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-1-methyltetrazole, 98%

Product Code. 11414407
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414407 5 g 5g
11424407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414407 Supplier Thermo Scientific Alfa Aesar Supplier No. B20443.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13183-79-4 | C2H4N4S | 116.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13183-79-4
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00037317
InChI Key XOHZHMUQBFJTNH-UHFFFAOYSA-N
Synonym 5-mercapto-1-methyltetrazole, 1-methyl-5-mercaptotetrazole, 1-methyl-1h-tetrazole-5-thiol, 1-n-methyl-5-thiotetrazole, 1-methyltetrazole-5-thiol, n-methyltetrazolethiol, 1-methyl-5-tetrazolethione, 1-methyl-5-mercapto-1h-tetrazole, 1-methyl-5-mercapto-1,2,3,4-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-methyl
PubChem CID 2723772
IUPAC Name 1-methyl-2H-tetrazole-5-thione
SMILES CN1NN=NC1=S

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Formula Weight 116.14
Percent Purity 98%
Chemical Name or Material 5-Mercapto-1-methyltetrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.