missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5 Lipoxygenase Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10021-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.Specifications
| 5 Lipoxygenase | |
| Polyclonal | |
| Unconjugated | |
| ALOX5 | |
| 5 Lipoxygenase, 5 LO, 5 LOX, 5LPG, ALOX5, LOG5 | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09917, P48999 | |
| ALOX5 | |
| Protein A | |
| RUO | |
| 11689, 240 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction